Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride, MP Biomedicals™

Catalog No. ICN15584625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15584625 25 g
ICN15584605 5 g
ICN15584610 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15584625 Supplier MP Biomedicals, Inc Supplier No. 0215584625
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 51298-62-5 | C7H16ClN5O4 | 269.69 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51298-62-5
Molecular Formula C7H16ClN5O4
Molecular Weight (g/mol) 269.69
MDL Number MFCD00039052,MFCD00133613
InChI Key QBNXAGZYLSRPJK-JEDNCBNOSA-N
Synonym h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome
PubChem CID 135193
IUPAC Name hydrogen methyl (2S)-2-amino-5-{[amino(nitroamino)methylidene]amino}pentanoate chloride
SMILES [H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 269.7
Grade Biochemical
Physical Form Powder
Chemical Name or Material N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride
EINECSNumber 257-116-1
Recommended Storage Store at -0°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.