Learn More
Medchemexpress LLC N-nitroso-3-azabicyclo[3.3.0]octane-d4 | 1346604-35-0 | 95.0% | 144.21 g/mol | C7H8D4N2O | 5 MG
SDP
Catalog No. 5000222594
Encompass_Preferred
N-Nitroso-3-azabicyclo[3.3.0]octane-d4 is the deuterium-labeled isotopologue of N-nitroso-3-azabicyclo[3.3.0]octane provided as a reference material for analytical and research applications. It is supplied as a solid for use as an internal standard in mass spectrometry and isotopic labeling studies; not for human or veterinary use.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for quantitative and qualitative analytical methods.
  • Provided with batch-specific certificate of analysis.
  • Stable solid form suitable for recommended long-term storage.
  • Intended for research use only.
Catalog No. 50-002-22594 Supplier Medchemexpress LLC Supplier No. HY143812S5MG
N-Nitroso-3-azabicyclo[3.3.0]octane-d4 is the deuterium-labeled isotopologue of N-nitroso-3-azabicyclo[3.3.0]octane provided as a reference material for analytical and research applications. It is supplied as a solid for use as an internal standard in mass spectrometry and isotopic labeling studies; not for human or veterinary use.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for quantitative and qualitative analytical methods.
  • Provided with batch-specific certificate of analysis.
  • Stable solid form suitable for recommended long-term storage.
  • Intended for research use only.