Learn More
Medchemexpress LLC N-Nitroso-3-azabicyclo[3.3.0]octane-d4 | 1346604-35-0 | 95.0% | C7H8D4N2O | 100 MG
SDP
Catalog No. 5000356155
Encompass_Preferred
N-Nitroso-3-azabicyclo[3.3.0]octane-d4 is the deuterium labeled N-Nitroso-3-azabicyclo[3.3.0]octane. It is for research use only and not intended for human use. Deuterium labeled compounds can be used as tracers or internal standards for quantitative analysis by methods like NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-56155 Supplier Medchemexpress LLC Supplier No. HY143812S100MG
N-Nitroso-3-azabicyclo[3.3.0]octane-d4 is the deuterium labeled N-Nitroso-3-azabicyclo[3.3.0]octane. It is for research use only and not intended for human use. Deuterium labeled compounds can be used as tracers or internal standards for quantitative analysis by methods like NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.