Learn More
Medchemexpress LLC N-nitroso-3-azabicyclo[3.3.0]octane-d4 | 1346604-35-0 | 95.0% | C7H8D4N2O | 10MG
SDP
Catalog No. 5000258410
Encompass_Preferred
N-Nitroso-3-azabicyclo[3.3.0]octane-d4 is a deuterium-labeled N-nitroso azabicyclic compound supplied for research use as an isotope-labeled standard. It is intended for use as a tracer or internal standard in quantitative analyses by NMR, GC-MS, or LC-MS, and is provided with specified purity and cold-storage recommendations to maintain stability.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Provided with stated purity of 95.0%.
  • Available in multiple small pack sizes, including 10 mg, for trace-level work.
  • Recommended storage: pure form at -20°C; in solvent, -80°C for extended stability.
Catalog No. 50-002-58410 Supplier Medchemexpress LLC Supplier No. HY143812S10MG
N-Nitroso-3-azabicyclo[3.3.0]octane-d4 is a deuterium-labeled N-nitroso azabicyclic compound supplied for research use as an isotope-labeled standard. It is intended for use as a tracer or internal standard in quantitative analyses by NMR, GC-MS, or LC-MS, and is provided with specified purity and cold-storage recommendations to maintain stability.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Provided with stated purity of 95.0%.
  • Available in multiple small pack sizes, including 10 mg, for trace-level work.
  • Recommended storage: pure form at -20°C; in solvent, -80°C for extended stability.