Learn More
Cayman Chemical N-NItroso Atenolol 10mg
SDP
Catalog No. 502910292
Encompass_Preferred
A genotoxic derivative of (±)-atenolol; induces DNA fragmentation in rat hepatocytes from 0.1 to 1 mM
Catalog No. 50-291-0292 Supplier Cayman Chemical Supplier No. 3473510 MG
A genotoxic derivative of (±)-atenolol; induces DNA fragmentation in rat hepatocytes from 0.1 to 1 mM