Learn More
Cayman Chemical N-Nitroso Fluoxetine, Purity: ≥95%, CAS: 150494-06-7, 25 mg
SDP
Catalog No. 502911274
Encompass_Preferred
A derivative of fluoxetine; genotoxic to S. typhimurium at 0.06-0.12 mg/ml
Catalog No. 50-291-1274 Supplier Cayman Chemical Supplier No. 3487925 MG
A derivative of fluoxetine; genotoxic to S. typhimurium at 0.06-0.12 mg/ml