Learn More
Cayman Chemical N-NItroso Metoprolol 10mg
SDP
Catalog No. 502910872
Encompass_Preferred
A genotoxic derivative of metoprolol; induces DNA fragmentation in rat hepatocytes from 0.1-1 mM; induces micronucleation of rat hepatocytes in vivo at 1,000 mg/kg
Catalog No. 50-291-0872 Supplier Cayman Chemical Supplier No. 3473810 MG
A genotoxic derivative of metoprolol; induces DNA fragmentation in rat hepatocytes from 0.1-1 mM; induces micronucleation of rat hepatocytes in vivo at 1,000 mg/kg