Learn More
Cayman Chemical N-NItroso-N-ethylnIlIn 10mg
SDP
Catalog No. 502983885
Encompass_Preferred
An N-nitrosamine; non-carcinogenic in a survival assay using 2FR450 Rauscher leukemia virus-infected rat embryo cultures; a synthetic intermediate in the synthesis of 3-amino-4-hydrazine-cyclobut-3-ene-1,2-diones as CXCR2 antagonists
Catalog No. 50-298-3885 Supplier Cayman Chemical Supplier No. 2106310 MG
An N-nitrosamine; non-carcinogenic in a survival assay using 2FR450 Rauscher leukemia virus-infected rat embryo cultures; a synthetic intermediate in the synthesis of 3-amino-4-hydrazine-cyclobut-3-ene-1,2-diones as CXCR2 antagonists