Learn More
Cayman Chemical N-Nitroso-N-ethylaniline, Purity: ≥98%, CAS: 612-64-6, 25 mg
SDP
Catalog No. 502982652
Encompass_Preferred
An N-nitrosamine; non-carcinogenic in a survival assay using 2FR450 Rauscher leukemia virus-infected rat embryo cultures; a synthetic intermediate in the synthesis of 3-amino-4-hydrazine-cyclobut-3-ene-1,2-diones as CXCR2 antagonists
Catalog No. 50-298-2652 Supplier Cayman Chemical Supplier No. 2106325 MG
An N-nitrosamine; non-carcinogenic in a survival assay using 2FR450 Rauscher leukemia virus-infected rat embryo cultures; a synthetic intermediate in the synthesis of 3-amino-4-hydrazine-cyclobut-3-ene-1,2-diones as CXCR2 antagonists