Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-nitroso-n-methyl-4-aminobutyric acid | 61445-55-4 | MFCD00869939 | 99.9% | 146.14 | C5H10N2O3 | 50 MG
SDP

Catalog No. 5000284552
Encompass_Preferred

N-nitroso-n-methyl-4-aminobutyric acid is a tobacco-derived nitrosamine used as a research chemical and analytical reference. It is supplied as a colorless to light yellow liquid with high reported purity and is intended for research use only.

  • High purity (HPLC): 99.88%.
  • Molecular formula C5H10N2O3.
  • Molecular weight 146.14.
  • Appearance liquid, colorless to light yellow.
  • Available in small milligram packs, including 50 mg.
  • CAS number 61445-55-4.
  • Tobacco-specific nitrosamine; identified as a carcinogen.
  • For research use only; not for sale to patients.

Catalog No. 50-002-84552 Supplier Medchemexpress LLC Supplier No. HYW67989150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-nitroso-n-methyl-4-aminobutyric acid is a tobacco-derived nitrosamine used as a research chemical and analytical reference. It is supplied as a colorless to light yellow liquid with high reported purity and is intended for research use only.

  • High purity (HPLC): 99.88%.
  • Molecular formula C5H10N2O3.
  • Molecular weight 146.14.
  • Appearance liquid, colorless to light yellow.
  • Available in small milligram packs, including 50 mg.
  • CAS number 61445-55-4.
  • Tobacco-specific nitrosamine; identified as a carcinogen.
  • For research use only; not for sale to patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.