Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-nitroso-N-methyl-4-aminobutyric acid | 61445-55-4 | MFCD00869939 | 99.9% | 146.14 g/mol | C5H10N2O3 | 100 MG
SDP

Catalog No. 5000283951
Encompass_Preferred

N-Nitroso-N-methyl-4-aminobutyric acid is a tobacco-specific nitrosamine provided as a high-purity research reagent for analytical and toxicological studies. It is intended for laboratory research use only and supplied as a solid; handle with appropriate carcinogen safety precautions.

  • Tobacco-specific nitrosamine and carcinogen.
  • Used as an analytical standard and in toxicology studies.
  • Molecular weight 146.14 g/mol.
  • Chemical formula C5H10N2O3.
  • CAS number 61445-55-4.
  • Reported purity 99.9%.
  • Supplied as a 100 mg solid.
  • Certificate of analysis available.

Catalog No. 50-002-83951 Supplier Medchemexpress LLC Supplier No. HYW679891100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Nitroso-N-methyl-4-aminobutyric acid is a tobacco-specific nitrosamine provided as a high-purity research reagent for analytical and toxicological studies. It is intended for laboratory research use only and supplied as a solid; handle with appropriate carcinogen safety precautions.

  • Tobacco-specific nitrosamine and carcinogen.
  • Used as an analytical standard and in toxicology studies.
  • Molecular weight 146.14 g/mol.
  • Chemical formula C5H10N2O3.
  • CAS number 61445-55-4.
  • Reported purity 99.9%.
  • Supplied as a 100 mg solid.
  • Certificate of analysis available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.