Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Nitroso-N-methylaniline 98.0+%, TCI America™
SDP

Catalog No. N026425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N026425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N026425ML Supplier TCI America Supplier No. N026425ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 614-00-6
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00045675
InChI Key MAXCWSIJKVASQC-UHFFFAOYSA-N
Synonym N-Methyl-N-nitrosoaniline, N-Methyl-N-phenylnitrosamine
PubChem CID 11957
IUPAC Name N-methyl-N-phenylnitrous amide
SMILES CN(C1=CC=CC=C1)N=O

Specifications

Boiling Point 128°C
Quantity 25 mL
UN Number 2810
Formula Weight 136.15
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material N-Nitroso-N-methylaniline
EINECSNumber (3)-0155
RTECSNumber BY5775000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.