Learn More
Cayman Chemical N-NItroso-N-methylnIlIn 250mg
SDP
Catalog No. 502910150
Encompass_Preferred
An N-nitrosamine; induces the formation of esophageal, pharyngeal, and tongue tumors in rats at 200 mg/L in the drinking water or 15 mg/ml via oral gavage; toxic to rats (LD50s = 336 and 225 mg/kg for male and female rats, respectively); reduces survival of hamsters but only induces a low incidence of tumors at 5 mg/week for a total dose of 1.8 mmol
Catalog No. 50-291-0150 Supplier Cayman Chemical Supplier No. 35563250 MG
An N-nitrosamine; induces the formation of esophageal, pharyngeal, and tongue tumors in rats at 200 mg/L in the drinking water or 15 mg/ml via oral gavage; toxic to rats (LD50s = 336 and 225 mg/kg for male and female rats, respectively); reduces survival of hamsters but only induces a low incidence of tumors at 5 mg/week for a total dose of 1.8 mmol