Learn More
Selleck Chemical LLC N-Nitroso-N-methylurea
SDP
Catalog No. 503138289
Encompass_Preferred
N-Nitroso-N-methylurea (NMU MNU NMH 1-Methyl-1-nitrosourea N-Methyl-Nnitrosourea Methylnitrosourea) is a highly reliable carcinogen mutagen and teratogen N-Nitroso-N-methylurea can be used to induce animal models of Breast Carcinoma
Catalog No. 50-313-8289 Supplier Selleck Chemical LLC Supplier No. E0158100MG
N-Nitroso-N-methylurea (NMU MNU NMH 1-Methyl-1-nitrosourea N-Methyl-Nnitrosourea Methylnitrosourea) is a highly reliable carcinogen mutagen and teratogen N-Nitroso-N-methylurea can be used to induce animal models of Breast Carcinoma