Learn More
Medchemexpress LLC HY-34758 100mg Medchemexpress, N-Nitroso-N-methylurea CAS:684-93-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY34758100MG
Encompass_Preferred
Medchemexpress, HY-34758 100mg N-Nitroso-N-methylurea CAS:684-93-5 N-Nitroso-N-methylurea (NMU;MNU;NMH) is a potent carcinogen, mutagen and teratogenand. N-Nitroso-N-methylurea is a direct-acting alkylating agent that interacts with DNA. N-Nitroso-N-methylurea targets multiple animal organs to cause various cancer and/or degenerative disease. N-Nitroso-N-methylurea is also a precursor in the synthesis of diazomethane[2][3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0930