Learn More
TARGETMOL CHEMICALS INC N-Nitroso-N-methylurea 500MG
SDP
Catalog No. 502879023
Encompass_Preferred
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. N-Nitroso-N-methylurea is a nitrosourea compound with alkylating, carcinogenic and mutagenic properties. It targets a variety of animal organs, causes various cancers and degenerative diseases, and can be used in diazomethane synthesis. Purity 99.89%
Catalog No. 50-287-9023 Supplier TARGETMOL CHEMICALS INC Supplier No. T4169500MG
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. N-Nitroso-N-methylurea is a nitrosourea compound with alkylating, carcinogenic and mutagenic properties. It targets a variety of animal organs, causes various cancers and degenerative diseases, and can be used in diazomethane synthesis. Purity 99.89%