Learn More
Medchemexpress LLC N-Nitroso-N-methylurea | 684-93-5 | 103.08 | 5 G
SDP
Catalog No. 5000404981
Encompass_Preferred
N-Nitroso-N-methylurea is a potent carcinogen, mutagen, and teratogen. It functions as a direct-acting alkylating agent, interacting with DNA, and is known to target multiple animal organs, potentially leading to various cancers or degenerative diseases. This compound also serves as a precursor in the synthesis of diazomethane. It is a flammable solid intended for laboratory use in the manufacture of substances and is stable under recommended storage conditions.
  • Functions as a direct-acting alkylating agent
  • Interacts with DNA
  • Targets multiple animal organs
  • Leads to various cancers and/or degenerative diseases
  • Precursor in the synthesis of diazomethane
  • Used in animal modeling to construct tumor models
Catalog No. 50-004-04981 Supplier Medchemexpress LLC Supplier No. HY347585G
N-Nitroso-N-methylurea is a potent carcinogen, mutagen, and teratogen. It functions as a direct-acting alkylating agent, interacting with DNA, and is known to target multiple animal organs, potentially leading to various cancers or degenerative diseases. This compound also serves as a precursor in the synthesis of diazomethane. It is a flammable solid intended for laboratory use in the manufacture of substances and is stable under recommended storage conditions.
  • Functions as a direct-acting alkylating agent
  • Interacts with DNA
  • Targets multiple animal organs
  • Leads to various cancers and/or degenerative diseases
  • Precursor in the synthesis of diazomethane
  • Used in animal modeling to construct tumor models