Learn More
Medchemexpress LLC N-Nitroso-N-methylurea | 684-93-5 | 99.7% | 103.08 | 500 MG
SDP
Catalog No. 5000404544
Encompass_Preferred
N-Nitroso-N-methylurea is a potent carcinogen, mutagen, and teratogen. It acts as a direct-acting alkylating agent that interacts with DNA, targeting multiple animal organs to induce various cancers and/or degenerative diseases. It is also utilized as a precursor in the synthesis of diazomethane.
  • Potent carcinogen, mutagen, and teratogen
  • Direct-acting alkylating agent that interacts with DNA
  • Targets multiple animal organs to induce various cancers and/or degenerative diseases
  • Precursor in the synthesis of diazomethane
  • Can be used to construct rat tumor models
  • Increases cellular NF-κB activity and I-κBα phosphorylation in human malignant keratinocytes
Catalog No. 50-004-04544 Supplier Medchemexpress LLC Supplier No. HY34758500MG
N-Nitroso-N-methylurea is a potent carcinogen, mutagen, and teratogen. It acts as a direct-acting alkylating agent that interacts with DNA, targeting multiple animal organs to induce various cancers and/or degenerative diseases. It is also utilized as a precursor in the synthesis of diazomethane.
  • Potent carcinogen, mutagen, and teratogen
  • Direct-acting alkylating agent that interacts with DNA
  • Targets multiple animal organs to induce various cancers and/or degenerative diseases
  • Precursor in the synthesis of diazomethane
  • Can be used to construct rat tumor models
  • Increases cellular NF-κB activity and I-κBα phosphorylation in human malignant keratinocytes