Learn More
Cayman Chemical N-NItroso SarcosIn 100mg
SDP
Catalog No. 502910258
Encompass_Preferred
A carcinogen; induces the formation of metastatic liver carcinomas in mice at 225 mg/kg
Catalog No. 50-291-0258 Supplier Cayman Chemical Supplier No. 34734100 MG
A carcinogen; induces the formation of metastatic liver carcinomas in mice at 225 mg/kg