Learn More
Cayman Chemical N-NItroso SarcosIn-d3 5mg
SDP
Catalog No. 502910587
Encompass_Preferred
An internal standard for the quantification of nitroso sarcosine by GC- or LC-MS
Catalog No. 50-291-0587 Supplier Cayman Chemical Supplier No. 347405 MG
An internal standard for the quantification of nitroso sarcosine by GC- or LC-MS