Learn More
Medchemexpress LLC N-Nitrosoanatabine-d4 | 1020719-69-0 | C10H7D4N3O | 500 UG
SDP
Catalog No. 5000355618
Encompass_Preferred
N-Nitrosoanatabine-d4 is the deuterium labeled N-Nitrosoanatabine. It can be used as a tracer or as an internal standard for quantitative analysis. Deuteration, the incorporation of stable heavy isotopes into drug molecules, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs. This compound is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-55618 Supplier Medchemexpress LLC Supplier No. HY145476S500UG
N-Nitrosoanatabine-d4 is the deuterium labeled N-Nitrosoanatabine. It can be used as a tracer or as an internal standard for quantitative analysis. Deuteration, the incorporation of stable heavy isotopes into drug molecules, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs. This compound is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs