Learn More
Medchemexpress LLC N-Nitrosoanatabine (N-NAT) | 887407-16-1 | 99.7% | C10H11N3O | 1 MG
SDP
Catalog No. 5000355617
Encompass_Preferred
(R,S)-N-Nitrosoanatabine (N-NAT) is a tobacco-specific nitrosamine (TSNA) found in tobacco, tobacco products, and electronic cigarettes.
  • The (S) enantiomer constitutes approximately 80% of N-NAT in commercial and research cigarettes and smokeless tobacco.
  • It is not carcinogenic in rats when administered at doses of 1, 3, and 9 mmol/kg.
Catalog No. 50-003-55617 Supplier Medchemexpress LLC Supplier No. HY1454761MG
(R,S)-N-Nitrosoanatabine (N-NAT) is a tobacco-specific nitrosamine (TSNA) found in tobacco, tobacco products, and electronic cigarettes.
  • The (S) enantiomer constitutes approximately 80% of N-NAT in commercial and research cigarettes and smokeless tobacco.
  • It is not carcinogenic in rats when administered at doses of 1, 3, and 9 mmol/kg.