Learn More
Cayman Chemical N-Nitrosodibutylamine, Purity: ≥95%, CAS: 924-16-3, 100 mg
SDP
Catalog No. 502909924
Encompass_Preferred
An N-nitrosamine; induces apoptosis and ROS production in HL-60 cells at 10 and 20 mM; induces tumor formation in rats at 0.05% in the drinking water; has been found in various food products, including fresh vegetables and pork, and influent and effluent of wastewater
Catalog No. 50-290-9924 Supplier Cayman Chemical Supplier No. 35474100 MG
An N-nitrosamine; induces apoptosis and ROS production in HL-60 cells at 10 and 20 mM; induces tumor formation in rats at 0.05% in the drinking water; has been found in various food products, including fresh vegetables and pork, and influent and effluent of wastewater