Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Nitrosodibutylamine | 924-16-3 | 100.0% | 1 ML
SDP

Catalog No. 5000340724
Encompass_Preferred

N-Nitrosodibutylamine is a light yellow to yellow liquid chemical (C8H18N2O, Molecular Weight: 158.24) that is found in drinking water. It is known to regulate the caspase pathway, induce apoptosis, and can cause hepatocarcinoma and oxidative DNA damage. This product is supplied as a 10 mM solution in 1 mL of DMSO and is intended for research use only.

  • Regulates caspase pathway
  • Induces apoptosis
  • Causes hepatocarcinoma
  • Induces oxidative DNA damage
  • Supplied as a 10 mM solution in 1 mL of DMSO
  • Appearance: Liquid
  • Color: Light yellow to yellow

Catalog No. 50-003-40724 Supplier Medchemexpress LLC Supplier No. HY13111310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Nitrosodibutylamine is a light yellow to yellow liquid chemical (C8H18N2O, Molecular Weight: 158.24) that is found in drinking water. It is known to regulate the caspase pathway, induce apoptosis, and can cause hepatocarcinoma and oxidative DNA damage. This product is supplied as a 10 mM solution in 1 mL of DMSO and is intended for research use only.

  • Regulates caspase pathway
  • Induces apoptosis
  • Causes hepatocarcinoma
  • Induces oxidative DNA damage
  • Supplied as a 10 mM solution in 1 mL of DMSO
  • Appearance: Liquid
  • Color: Light yellow to yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.