Learn More
Medchemexpress LLC N-Nitrosodibutylamine | 924-16-3 | 100.0% | 250 MG
SDP
Catalog No. 5000341474
Encompass_Preferred
N-Nitrosodibutylamine is a nitrosamine found in drinking water. It regulates the caspase pathway, induces apoptosis, and causes hepatocarcinoma and oxidative DNA damage. It is for research use only.
  • Regulates the caspase pathway
  • Induces apoptosis
  • Causes hepatocarcinoma and oxidative DNA damage
  • For research use only
Catalog No. 50-003-41474 Supplier Medchemexpress LLC Supplier No. 250MGHY131113
N-Nitrosodibutylamine is a nitrosamine found in drinking water. It regulates the caspase pathway, induces apoptosis, and causes hepatocarcinoma and oxidative DNA damage. It is for research use only.
  • Regulates the caspase pathway
  • Induces apoptosis
  • Causes hepatocarcinoma and oxidative DNA damage
  • For research use only