Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-nitrosodicyclohexylamine | 947-92-2 | MFCD00021331 | 99.0% | 210.32 g/mol | C12H22N2O | 1 ML
SDP

Catalog No. 5000283914
Encompass_Preferred

N-Nitrosodicyclohexylamine (NDCHA) is provided as a 10 mM solution in dimethyl sulfoxide (DMSO) for research use. It is reported to have anti-androgenic activity and is supplied at high purity for in vitro pharmacology and biochemical studies.

  • 10 mM solution in DMSO.
  • Packaged as a 1 mL vial.
  • Purity ≥99.0%.
  • Molecular weight 210.32 g/mol.
  • For research use only; not for human use.

Catalog No. 50-002-83914 Supplier Medchemexpress LLC Supplier No. HYW65372310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Nitrosodicyclohexylamine (NDCHA) is provided as a 10 mM solution in dimethyl sulfoxide (DMSO) for research use. It is reported to have anti-androgenic activity and is supplied at high purity for in vitro pharmacology and biochemical studies.

  • 10 mM solution in DMSO.
  • Packaged as a 1 mL vial.
  • Purity ≥99.0%.
  • Molecular weight 210.32 g/mol.
  • For research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.