Learn More
Medchemexpress LLC HY-N7434 500mg Medchemexpress, N-Nitrosodiethylamine CAS:55-18-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN7434500MG
Encompass_Preferred
Medchemexpress, HY-N7434 500mg N-Nitrosodiethylamine CAS:55-18-5 N-Nitrosodiethylamine is a potent hepatocarcinogenic dialkylnitrosoamine. N-Nitrosodiethylamine is mainly present in tobacco smoke, water, cheddar cheese, cured, fried meals and many alcoholic beverages. N-Nitrosodiethylamine is responsible for the changes in the nuclear enzymes associated with DNA repair/replication. N-Nitrosodiethylamine results in various tumors in all animal species. The main target organs are the nasal cavity, trachea, lung, esophagus and liver. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2998