Learn More
Medchemexpress LLC N-Nitrosodiethylamine-d10 | 1219794-54-3 | 1 MG
SDP
Catalog No. 5000418051
Encompass_Preferred
N-Nitrosodiethylamine-d10 is the deuterium labeled N-Nitrosodiethylamine, a potent hepatocarcinogenic dialkylnitrosoamine found in various food products and tobacco smoke. It impacts nuclear enzymes related to DNA repair and replication, leading to tumors primarily in the nasal cavity, trachea, lung, esophagus, and liver. Deuterium labeling incorporates stable heavy isotopes into drug molecules, serving as tracers for quantitation during drug development and potentially influencing pharmacokinetic and metabolic profiles.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
  • Affects DNA/RNA synthesis.
  • Involved in cell cycle/DNA damage pathways.
Catalog No. 50-004-18051 Supplier Medchemexpress LLC Supplier No. HYN7434S11MG
N-Nitrosodiethylamine-d10 is the deuterium labeled N-Nitrosodiethylamine, a potent hepatocarcinogenic dialkylnitrosoamine found in various food products and tobacco smoke. It impacts nuclear enzymes related to DNA repair and replication, leading to tumors primarily in the nasal cavity, trachea, lung, esophagus, and liver. Deuterium labeling incorporates stable heavy isotopes into drug molecules, serving as tracers for quantitation during drug development and potentially influencing pharmacokinetic and metabolic profiles.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
  • Affects DNA/RNA synthesis.
  • Involved in cell cycle/DNA damage pathways.