Learn More
Selleck Chemical LLC N-Nitrosodiethylamine-E0136-1G
SDP
Catalog No. 503138079
Encompass_Preferred
N-Nitrosodiethylamine (N-Nitroso-diethylamine NDEA diethylnitrosamine DEN) is DNA reactive after bioactivation and produces tumors in many animal species N-Nitrosodiethylamine can be used to induce animal models of Lung Carcinoma
Catalog No. 50-313-8079 Supplier Selleck Chemical LLC Supplier No. E01361G
N-Nitrosodiethylamine (N-Nitroso-diethylamine NDEA diethylnitrosamine DEN) is DNA reactive after bioactivation and produces tumors in many animal species N-Nitrosodiethylamine can be used to induce animal models of Lung Carcinoma