Learn More
Selleck Chemical LLC N-Nitrosodiethylamine-E0136-5G
SDP
Catalog No. 503138334
Encompass_Preferred
N-Nitrosodiethylamine (N-Nitroso-diethylamine NDEA diethylnitrosamine DEN) is DNA reactive after bioactivation and produces tumors in many animal species N-Nitrosodiethylamine can be used to induce animal models of Lung Carcinoma
Catalog No. 50-313-8334 Supplier Selleck Chemical LLC Supplier No. E01365G
N-Nitrosodiethylamine (N-Nitroso-diethylamine NDEA diethylnitrosamine DEN) is DNA reactive after bioactivation and produces tumors in many animal species N-Nitrosodiethylamine can be used to induce animal models of Lung Carcinoma