Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Nitrosodiethylamine, MilliporeSigma™ Supelco™

Catalog No. 111018995 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8995 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8995 Supplier MilliporeSigma Supelco Supplier No. 442687
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 55-18-5
Density 0.95 g/mL
Physical Form Neat
Linear Formula (C2H5)2 NNO
MDL Number MFCD00013890
Boiling Point 177°C
Synonym Diethylnitrosamine
Type Food and Cosmetic Component Standard
Quantity 1000 mg
Formula Weight 102.14
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.