Learn More
Sigma Aldrich Fine Chemicals Biosciences N-Nitrosodiethylamine Phar1G
Catalog No. 502902139
Encompass
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Catalog No. 50-290-2139 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PHR24081G
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards