Learn More
Chem Service Inc N-Nitrosodimethylamine Solution, 62-75-9, MFCD00002053, 5 mL
SDP
Supplier:  Chem Service Inc S12572M15ML
Encompass_Preferred
Molecular Formula: C2-H6-N2-O, MW: 74.08, 99.5%, Alternate Name(s): NDMA. Concentration and Solvent: 100 ug/ml in Methanol.
Catalog No. NC9391357