Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Nitrosodimethylamine, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023033 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-3033 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-3033 Supplier MilliporeSigma Supelco Supplier No. PHR2407100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

TRUSTED_SUSTAINABILITY

Specifications

CAS 62-75-9
Vapor Pressure 5 mmHg (20°C)
Density 1.01 g/mL (literature)
Flash Point 61°C (Closed Cup)
Molecular Formula C2H6N2O
Linear Formula (CH3)2 NNO
MDL Number MFCD00002053
Boiling Point 153°C (774 mmHg) (literature)
Synonym N-Nitrosodimethylamine; Dimethylnitrosamine
Quantity 100 mg
Formula Weight 74.08
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.