Learn More
Medchemexpress LLC N-Nitrosodipropylamine-d14 | 93951-96-3 | 98.5% | 144.27 | 1 MG
SDP
Catalog No. 5000404099
Encompass_Preferred
N-Nitrosodipropylamine-d14 is the deuterium labeled N-Nitrosodipropylamine. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-04099 Supplier Medchemexpress LLC Supplier No. HY24385S1MG
N-Nitrosodipropylamine-d14 is the deuterium labeled N-Nitrosodipropylamine. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.