Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Nitrosodipropylamine 98.0+%, TCI America™
SDP

Catalog No. N044425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N044425G 25 g
N04445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N044425G Supplier TCI America Supplier No. N044425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 621-64-7
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.191
MDL Number MFCD00013891
InChI Key YLKFDHTUAUWZPQ-UHFFFAOYSA-N
Synonym n-nitrosodipropylamine, n-nitrosodi-n-propylamine, dipropylnitrosamine, n,n-dipropylnitrosamine, 1-propanamine, n-nitroso-n-propyl, nitrosodipropylamine, dipropylamine, n-nitroso, di-n-propylnitrosamine, di-n-propylnitrosoamine, n-nitroso-n-propyl-1-propanamine
PubChem CID 12130
ChEBI CHEBI:82358
IUPAC Name N,N-dipropylnitrous amide
SMILES CCCN(CCC)N=O

Specifications

Color Yellow
Boiling Point 113°C
Quantity 25 g
UN Number 3082
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Nitrosodipropylamine
RTECSNumber JL9700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.