Learn More
Medchemexpress LLC N-Nitrosomorpholine | 59-89-2 | 99.98% | 5 G
SDP
Catalog No. 5000339918
Encompass_Preferred
N-Nitrosomorpholine is a photosensitive nitrosamine that is carcinogenic and can be used to construct animal cancer models. It is for research use only and not sold to patients.
  • Photosensitive nitrosamine
  • Carcinogenic agent
  • Used to construct animal cancer models
  • For research use only
  • Not sold to patients
Catalog No. 50-003-39918 Supplier Medchemexpress LLC Supplier No. HY1311235G
N-Nitrosomorpholine is a photosensitive nitrosamine that is carcinogenic and can be used to construct animal cancer models. It is for research use only and not sold to patients.
  • Photosensitive nitrosamine
  • Carcinogenic agent
  • Used to construct animal cancer models
  • For research use only
  • Not sold to patients