Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-nitrosomorpholine-d8 | 1219805-76-1 | 99.7% | C4D8N2O2 | 10MG
SDP

Catalog No. 5000261721
Encompass_Preferred

N-Nitrosomorpholine-d8 is the deuterium-labeled analog of N-nitrosomorpholine, provided as an isotope-labeled internal standard for analytical and tracer applications. It is intended for quantitative analysis and method development in NMR, GC-MS, and LC-MS assays. The compound is light-sensitive and is a known strong animal carcinogen; handle with appropriate safety precautions.

  • Deuterium labeled for use as an internal standard in quantitative NMR, GC-MS, and LC-MS.
  • High chemical purity (99.74%).
  • Molecular formula C4D8N2O2 and molecular weight 124.17.
  • Available in small, analytical sizes suitable for reference standards.
  • Light-sensitive; requires controlled storage and handling.
  • Suitable as a tracer for method development and quantitative analysis.

Catalog No. 50-002-61721 Supplier Medchemexpress LLC Supplier No. HY131123S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Nitrosomorpholine-d8 is the deuterium-labeled analog of N-nitrosomorpholine, provided as an isotope-labeled internal standard for analytical and tracer applications. It is intended for quantitative analysis and method development in NMR, GC-MS, and LC-MS assays. The compound is light-sensitive and is a known strong animal carcinogen; handle with appropriate safety precautions.

  • Deuterium labeled for use as an internal standard in quantitative NMR, GC-MS, and LC-MS.
  • High chemical purity (99.74%).
  • Molecular formula C4D8N2O2 and molecular weight 124.17.
  • Available in small, analytical sizes suitable for reference standards.
  • Light-sensitive; requires controlled storage and handling.
  • Suitable as a tracer for method development and quantitative analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.