Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-nitrosomorpholine-d8 | 1219805-76-1 | MFCD08704880 | 99.7% | 124.17 g·mol⁻1 | C4D8N2O2 | 5 MG
SDP

Catalog No. 5000262943
Encompass_Preferred

N-Nitrosomorpholine-d8 is a deuterium-labeled analogue of N-nitrosomorpholine used as an isotope-labeled reference standard and internal standard for analytical testing and quantitation of nitrosamines. It is photosensitive and a potent animal carcinogen; handle with appropriate engineering controls and personal protective equipment, and consult the SDS and batch COA for safety and purity details.

  • CAS number: 1219805-76-1.
  • Molecular formula: C4D8N2O2.
  • Molecular weight: 124.17 g·mol⁻1.
  • Purity (HPLC): 99.7%.
  • Appearance: Colorless to light yellow liquid.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended use: isotope-labeled reference standard for analytical and quantitation applications.

Catalog No. 50-002-62943 Supplier Medchemexpress LLC Supplier No. HY131123S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Nitrosomorpholine-d8 is a deuterium-labeled analogue of N-nitrosomorpholine used as an isotope-labeled reference standard and internal standard for analytical testing and quantitation of nitrosamines. It is photosensitive and a potent animal carcinogen; handle with appropriate engineering controls and personal protective equipment, and consult the SDS and batch COA for safety and purity details.

  • CAS number: 1219805-76-1.
  • Molecular formula: C4D8N2O2.
  • Molecular weight: 124.17 g·mol⁻1.
  • Purity (HPLC): 99.7%.
  • Appearance: Colorless to light yellow liquid.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended use: isotope-labeled reference standard for analytical and quantitation applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.