Learn More
AdipoGen N-Nitrosomorpholine NMOR
SDP
Supplier:  AdipoGen CDXN0370G001
Encompass_Preferred
Chemical. CAS 59-89-2. Formula C4H8N2O2. MW 116.12. N-Nitrosomorpholine NMOR is a nitrosamine and is known to be a potent carcinogen associated with cancer development in animals and humans. NMOR can damage DNA and cause mutations in genes that control cell growth and division. It reacts with DNA in vitro to form genotoxic activity, which may lead to cell death or mutagenesis. NMOR is formed in various industrial processes, including the synthesis of certain chemicals and in the preservation of cosmetic and personal care products. This compound can be used as analytical reference material. NMOR is used as a tumor initiator. It is a reagent for preparing carcinogenic animal diseases model. NMOR has other widespread utilization in laboratory and industrial environments. NMOR exhibits potent oxidizing properties, making it valuable in various applications, including as a bulding block for synthesis, environmental monitoring, and as a reagent in laboratory settings.
Catalog No. 50-256-0361