Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Nitrosomorpholine 99.0+%, TCI America™
SDP

Catalog No. N046625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N046625G 25 g
N04665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N046625G Supplier TCI America Supplier No. N046625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59-89-2
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00039710
InChI Key ZKXDGKXYMTYWTB-UHFFFAOYSA-N
Synonym n-nitrosomorpholine, nitrosomorpholine, morpholine, 4-nitroso, n-nitrosomorpholin, nmor, morpholine, n-nitroso, n-nitrosomorfolin, n-nitrosomorfolin czech, n-nitrosomorpholin german, unii-3l25fo7fn7
PubChem CID 6046
ChEBI CHEBI:76326
IUPAC Name 4-nitrosomorpholine
SMILES C1COCCN1N=O

Specifications

Color Yellow
Boiling Point 225°C
Quantity 25 g
UN Number 2811
Formula Weight 116.12
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material N-Nitrosomorpholine
RTECSNumber QE7525000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.