Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (±)-N′-Nitrosonornicotine-2,4,5,6-d4 | 66148-19-4 | MFCD01074209 | 99.1% | 181.23 | C9H7D4N3O | 1 MG
SDP

Catalog No. 5000281930
Encompass_Preferred

Deuterium-labeled N′-nitrosonornicotine (d4) is a stable-isotope reference standard intended for use as an internal standard in analytical and toxicology research. Supplied as a high-purity analytical reagent, it supports method development, calibration, and metabolite identification for GC-MS and LC-MS workflows.

  • Deuterium-labeled stable isotope internal standard.
  • High purity suitable for analytical use (≈99.1%).
  • Molecular weight 181.23 and formula C9H7D4N3O.
  • Optimized for GC-MS and LC-MS quantification.
  • Available in small research pack sizes, such as 1 mg.

Catalog No. 50-002-81930 Supplier Medchemexpress LLC Supplier No. HYW189682S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled N′-nitrosonornicotine (d4) is a stable-isotope reference standard intended for use as an internal standard in analytical and toxicology research. Supplied as a high-purity analytical reagent, it supports method development, calibration, and metabolite identification for GC-MS and LC-MS workflows.

  • Deuterium-labeled stable isotope internal standard.
  • High purity suitable for analytical use (≈99.1%).
  • Molecular weight 181.23 and formula C9H7D4N3O.
  • Optimized for GC-MS and LC-MS quantification.
  • Available in small research pack sizes, such as 1 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.