Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Nitrosonornicotine, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019748 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9748 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9748 Supplier MilliporeSigma Supelco Supplier No. 752852MG
Add to Cart
Add to Cart

N′-Nitrosonornicotine is a tobacco specific nitrosamine, which is found to be carcinogenic in nature. It is produced via nitrosation of nicotine during the process of curing, aging, processing and smoking of tobacco.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 80508-23-2
Physical Form Neat
Linear Formula C9H11N3 O
MDL Number MFCD00274581
Synonym 1-Nitroso-2-(3-pyridyl)pyrrolidine; 3-(1-Nitroso-2-pyrrolidinyl)pyridine; NNN
Type Analytical Standard
Quantity 2 mg
Formula Weight 177.20
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.