Learn More
Cayman Chemical N-NItrosonrnIcotIn 50mg
SDP
Catalog No. 502950378
Encompass_Preferred
An N-nitrosamine and a carcinogen; induces the formation of DNA adducts in isolated rat nasal mucosa and esophagus; induces tumor formation in rat esophagus and nasal cavity, mouse lung, forestomach, and trachea, and hamster trachea and forebrain; urinary levels are positively correlated with the risk of esophageal cancer in smokers
Catalog No. 50-295-0378 Supplier Cayman Chemical Supplier No. 3412050 MG
An N-nitrosamine and a carcinogen; induces the formation of DNA adducts in isolated rat nasal mucosa and esophagus; induces tumor formation in rat esophagus and nasal cavity, mouse lung, forestomach, and trachea, and hamster trachea and forebrain; urinary levels are positively correlated with the risk of esophageal cancer in smokers