Learn More
SPEX CERTIPREP LLC 3P N-NO2 CRM, 500 mL
Catalog No. NC0436408
Encompass_Preferred
(NSI by Zeptometrix) 1000mg/L in Water. Derived from high purity Sodium nitrite, NIST traceable.
Catalog No. NC0436408 Supplier SPEX CERTIPREP LLC 3P Supplier No. IS010500
(NSI by Zeptometrix) 1000mg/L in Water. Derived from high purity Sodium nitrite, NIST traceable.