Learn More
Medchemexpress LLC n-Nonadecane-d40 | 39756-36-0 | 100 MG
SDP
Catalog No. 5000419114
Encompass_Preferred
n-Nonadecane-d40 is a deuterium labeled Nonadecane intended for research use only. Deuteration, the incorporation of stable heavy isotopes of hydrogen (deuterium) into drug molecules, has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Utilized as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for methods such as NMR, GC-MS, or LC-MS
Catalog No. 50-004-19114 Supplier Medchemexpress LLC Supplier No. HYN7928S100MG
n-Nonadecane-d40 is a deuterium labeled Nonadecane intended for research use only. Deuteration, the incorporation of stable heavy isotopes of hydrogen (deuterium) into drug molecules, has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Utilized as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for methods such as NMR, GC-MS, or LC-MS