Learn More
Medchemexpress LLC n-Nonadecane-d40 | 39756-36-0 | 250 MG
SDP
Catalog No. 5000421954
Encompass_Preferred
n-Nonadecane-d40 is a deuterium labeled nonadecane, used for research purposes. It can serve as a tracer or an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS. Stable heavy isotopes like deuterium are incorporated into drug molecules as tracers for quantitation during drug development, which can affect pharmacokinetic and metabolic profiles.
  • Deuterium labeled compound
  • Suitable for research applications
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, and LC-MS
  • High purity of 99.2%
  • Isotopic enrichment of 99.4%
Catalog No. 50-004-21954 Supplier Medchemexpress LLC Supplier No. 250MGHYN7928S
Add to Cart
Add to Cart
n-Nonadecane-d40 is a deuterium labeled nonadecane, used for research purposes. It can serve as a tracer or an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS. Stable heavy isotopes like deuterium are incorporated into drug molecules as tracers for quantitation during drug development, which can affect pharmacokinetic and metabolic profiles.
  • Deuterium labeled compound
  • Suitable for research applications
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, and LC-MS
  • High purity of 99.2%
  • Isotopic enrichment of 99.4%