Learn More
Medchemexpress LLC N-Nonanoylglycine-d2 | 3008541-78-1 | ≥95.0% | 217.30 | C11H19D2NO3 | 10 MG
SDP
Catalog No. 5000288191
Encompass_Preferred
N-Nonanoylglycine-d2 is a deuterium-labeled analogue of N-nonanoylglycine intended for use as a stable isotope internal standard in analytical workflows such as liquid chromatography-mass spectrometry. It is supplied as a powdered reference material suitable for calibration, quantitation, and method validation in research and analytical laboratories.
  • Stable isotope-labeled internal standard for quantitative mass spectrometry.
  • Deuterium (d2) label provides a clear mass shift for selective detection and quantitation.
  • High purity suitable for analytical reference and calibration.
  • Available in small mass quantities for trace-level and method development work.
  • Supplied as a powder for straightforward storage and handling.
Catalog No. 50-002-88191 Supplier Medchemexpress LLC Supplier No. HY158084S10MG
N-Nonanoylglycine-d2 is a deuterium-labeled analogue of N-nonanoylglycine intended for use as a stable isotope internal standard in analytical workflows such as liquid chromatography-mass spectrometry. It is supplied as a powdered reference material suitable for calibration, quantitation, and method validation in research and analytical laboratories.
  • Stable isotope-labeled internal standard for quantitative mass spectrometry.
  • Deuterium (d2) label provides a clear mass shift for selective detection and quantitation.
  • High purity suitable for analytical reference and calibration.
  • Available in small mass quantities for trace-level and method development work.
  • Supplied as a powder for straightforward storage and handling.