Learn More
Dojindo Molecular Technologies Inc N-NONYL-B-D-THIOMALTOSIDE (1G), CAS: 148565-55-3, Purity: 98.0% (GC), MW: 484.60, C21H40O10S, suitable for isolating proteins inside of cell membranes
SDP
Catalog No. NC1778578
Encompass
n-Nonyl-β-D-thiomaltoside is a newly developed detergent whose lipophilic moiety is maltose. This reagent is suitable for isolating proteins inside of cell membranes. The activity of proteins isolated using n-Nonyl-β-D-thiomaltoside is higher than those isolated using n-Octyl-β-Dthioglucoside or n-Heptyl-β-D-thioglucoside. As the CMC value of this detergent is 2.4 mM, it is easy to remove from protein solutions by dialysis. Dojindo has various detergents for life science research. (https://www.dojindo.com/biochemicals/)
Catalog No. NC1778578 Supplier Dojindo Molecular Technologies Inc Supplier No. N37310
n-Nonyl-β-D-thiomaltoside is a newly developed detergent whose lipophilic moiety is maltose. This reagent is suitable for isolating proteins inside of cell membranes. The activity of proteins isolated using n-Nonyl-β-D-thiomaltoside is higher than those isolated using n-Octyl-β-Dthioglucoside or n-Heptyl-β-D-thioglucoside. As the CMC value of this detergent is 2.4 mM, it is easy to remove from protein solutions by dialysis. Dojindo has various detergents for life science research. (https://www.dojindo.com/biochemicals/)