Learn More
Cayman Chemical n-Nonyl-β-D-glucopyranoside, Purity: ≥95%, CAS: 69984-73-2, 1 g
SDP
Catalog No. 502935858
Encompass_Preferred
An anionic chiral surfactant; commonly used for the solubilization and crystallization of biological membrane proteins; has been used in the separation of drug enantiomers by micellar electrokinetic chromatography
Catalog No. 50-293-5858 Supplier Cayman Chemical Supplier No. 257051 G
An anionic chiral surfactant; commonly used for the solubilization and crystallization of biological membrane proteins; has been used in the separation of drug enantiomers by micellar electrokinetic chromatography